Anti-Ly6G/Ly6C (Gr-1) Rat Monoclonal Antibody [clone: RB6-8C5]

Supplier: BioXCell
BE0075-50MG BE0075-25MG BE0075-25MG BE0075-5MG BE0075-1MG BE0075-5MG BE0075-1MG BE0075-100MG BE0075-100MG BE0075-50MG
76556-860EA 3126.56 USD
76556-860 103572-768 103252-506 103252-508 76555-374 103252-502 76556-282 103252-504 76554-476 76557-150
Anti-Ly6G/Ly6C (Gr-1) Rat Monoclonal Antibody [clone: RB6-8C5]
Antibodies
InVivoMab anti-mouse Ly6G/Ly6C (Gr-1) , Clone: RB6-8C5, Isotype: rat IgG2b, κ, Applications: in vivo depletion of Gr-1+ myeloid cells, Flow cytometry, Immunohistochemistry (paraffin), Immunohistochemistry (frozen)

Type: Primary
Antigen: Ly6G/Ly6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR