Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ly6G Rat Monoclonal Antibody [clone: 1A8]

Supplier: Bio X Cell
BE0075-1-R025MG BE0075-1-R001MG BE0075-1-R100MG BE0075-1-R050MG BE0075-1-R005MG
103252-498 103572-766 103252-494 103252-500 103252-496
Anti-Ly6G Rat Monoclonal Antibody [clone: 1A8]
InVivoMab anti-mouse Ly6G, Clone: 1A8, Isotype: rat IgG2a, κ, Applications: in vivo neutrophil depletion, in vivo MDSC depletion, Immunofluorescence, Immunohistochemistry (paraffin), Immunohistochemistry (frozen), Flow cytometry

Type: Primary
Antigen: Ly6G
Clonality: Monoclonal
Clone: 1A8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty