Cyclohexanone

Danger

SR055ZA-204
BJSR055ZA-204EA 12029.3 USD
BJSR055ZA-204
Cyclohexanone
Cyclohexanone
Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155,6 °C
Melting Pt: -31 °C
Density: 0,95 g/cm3 (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
UN: 1915
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/12/57/771257.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR