Anti-CD8 alpha Mouse Monoclonal Antibody [clone: OKT-8]

Supplier: BioXCell
BE0004-2-50MG BE0004-2-1MG BE0004-2-5MG BE0004-2-5MG BE0004-2-100MG BE0004-2-100MG BE0004-2-25MG BE0004-2-50MG BE0004-2-25MG BE0004-2-1MG
76555-314EA 169.55 USD
76555-314 103252-210 76555-796 103572-406 76555-188 76555-562 103252-208 76556-490 103252-206 103252-212
Anti-CD8 alpha Mouse Monoclonal Antibody [clone: OKT-8]
Antibodies
InVivoMab anti-human CD8α, Clone: OKT-8, Isotype: mouse IgG2a, Applications: in vivo CD8+ T cell depletion in humanized mice

Type: Primary
Antigen: CD8alpha
Clonality: Monoclonal
Clone: OKT-8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR