L.D. Egg Yolk Agar Base Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M744-500G M744-500G
95022-212EA 495.05 USD
CA95022-212 95022-212
L.D. Egg Yolk Agar Base Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For detecting lecithinase activity of anaerobic microorganisms.
  • Microbiological culture media
  • 500 g poly bottles