Sobag Agar Plates, Teknova
Supplier: Teknova
S1011 S1010 S1010 S1012 S1012 S1011
101320-522PK 77.36 USD
101320-522 CA101320-522 CA100219-354 CA100217-694 100217-694 100219-354
Sobag Agar Plates, Teknova
Media for Microbiology Prepared Media for Microbiology
Sobag is similar to SOB (and SOC) with the addition of Ampicillin and Glucose.
It contains 2.0% Tryptone, 0.5% Yeast Extract, 0.05% Sodium Chloride, 10 mM Magnesium Chloride, 2% Glucose, 100 µg/mL Ampicillin and 100 µg/mL Nalidixic Acid or 1.5% Agar.