D-(-)-Luciferin potassium salt, StayBrite™
Supplier: BioVision
Synonyms:
Beetle Luciferin potassium salt
, Luciferin potassium salt
, Firefly luciferin potassium salt
, (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt
, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
D-Luciferin potassium salt is a useful substrate for the enzyme Firefly Luciferase. Upon oxidation by the enzyme luciferase, produces bioluminescence. D-Luciferin can be utilized to assay the expression of the luciferase gene linked to a promoter of interest. Alternatively, D-Luciferin and luciferase can be used to assess ATP availability in cellular or biochemical assays.
Storage Temperature: -20°C
Formula:
C₁₁H₇KN₂O₃S₂ MW: 318.41 g/mol |
MDL Number:
MFCD00044928 CAS Number: 115144-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...