1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol

Synonyms: 1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(phenylamino)propan-2-ol

2849-10 2849-50
76021-790EA 157.38 USD
76021-790 76021-792
1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol
1-(3,6-Dibromocarbazol-9-yl)-3-phenylaminopropan-2-ol

P7C3 is an aminopropyl carbazole derivative that acts as a proneurogenic, neuroprotective compound. Protects newborn neurons in the dentate gyrus by mitigating cell death. Also enhances learning and memory in aged rats. Orally available and brain-permeable.


Storage Temperature: -20°C

Formula: C₂₁H₁₈Br₂N₂O
MW: 474.21 g/mol
MDL Number: MFCD00572918
CAS Number: 301353-96-8
https://us.vwr.com/stibo/web/std.lang.all/23/17/23492317.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR