ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Synonyms: Nadide phosphate disodium saltNADP-Na2Nicotinamide-adenine dinucleotide phosphate disodium saltNADP disodium saltβ-NADH phosphate disodium saltβ-NADP-Na2

2736-1000 2736-100 2736-500
76021-610EA 788.06 USD
76021-610 76021-606 76021-608
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

NADP⁺ is a coenzyme that is widely distributed in living matter. It serves as an electron carrier in a number of reactions, being alternately oxidized (NADP+) and reduced (NADPH).


Storage Temperature: -20°C

Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37 g/mol
MDL Number: MFCD00065390
CAS Number: 24292-60-2
https://us.vwr.com/stibo/web/std.lang.all/22/54/23492254.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR