β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form)

2735-1000 2735-5000 2735-500
76021-600EA 99.24 USD
76021-600 76021-602 76021-604
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)

NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, beta-oxidation and the citric acid cycle. As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.


Storage Temperature: -20°C

Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MW: 709.41 g/mol
MDL Number: MFCD00036200
CAS Number: 606-68-8
https://us.vwr.com/stibo/web/std.lang.all/22/53/23492253.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR