β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form) ≥98% (by HPLC)

Synonyms: Nadide Nicotinamide adenine dinucleotide, oxidized NAD+

2734-500 2734-1000
76021-596EA 81.49 USD
76021-596 76021-598
β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form) ≥98% (by HPLC)
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)

NAD⁺ is a major electron acceptor molecule in biological oxidations. Acts as a coenzyme for hydride-transfer enzymes and a substrate for NAD(+)-consuming enzymes, which include ADP-ribose transferases, poly(ADP-ribose) polymerases, cADP-ribose synthases and sirtuins. Recently reported to be a P2Y11 agonist.


Storage Temperature: -20°C

Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
MDL Number: MFCD00036253
CAS Number: 53-84-9
https://us.vwr.com/stibo/web/std.lang.all/22/52/23492252.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR