D-alpha-Tocopherol succinate

Synonyms: Vitamin E Succinate

2722-50 2722-250
76021-544EA 262.69 USD
76021-544 76021-542
D-alpha-Tocopherol succinate
D-alpha-Tocopherol succinate

α-Tocopheryl succinate is an analog of α-Tocopherol (Vitamin E) with unique biological properties. It is the most effective form of Vitamin E in inducing differentiation, inhibition of proliferation and apoptosis in cancer cells. It does not affect the proliferation of most normal cells.


Storage Temperature: -20°C

Formula: C₃₃H₅₄O₅
MW: 530.79 g/mol
MDL Number: MFCD00072055
CAS Number: 4345-03-3
https://us.vwr.com/stibo/web/std.lang.all/22/40/23492240.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR