Kaempferol

Danger

Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

2523-250 2523-50
76020-826EA 391.11 USD
76020-826 76020-824
Kaempferol
Kaempferol

A naturally occurring polyphenol antioxidant that displays a wide range of pharmacological activities, including antioxidant, anti-inflammatory, antimicrobial, anticancer, cardioprotective, neuroprotective, antidiabetic, anti-osteoporotic, estrogenic/antiestrogenic, anxiolytic, analgesic and antiallergic activities. It reduces cell cycle progression in HT-29 cells. Kaempferol increases chromatin condensation, DNA fragmentation and the number of early apoptotic cells in HT-29 cells in a dose-dependent manner. In addition, kaempferol increases the levels of cleaved caspase-9, caspase-3 and caspase-7 as well as those of cleaved poly (ADP-ribose) polymerase. Moreover, it increases mitochondrial membrane permeability and cytosolic cytochrome c concentrations.


Storage Temperature: -20°C

Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00016938
CAS Number: 520-18-3
https://us.vwr.com/stibo/web/std.lang.all/20/47/23492047.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR