PF-03394197

Synonyms: N-Methyl-1-(trans-4-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexyl)methanesulfonamideN-Methyl-1-(trans-4-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexyl)methanesulphonamide

2883-25 2883-5
76021-880EA 169.28 USD
76021-880 76021-882
PF-03394197
PF-03394197

PF-03394197 is potent and selective Janus kinase (JAK) inhibitor


It inhibits JAK family members with an IC₅₀ of 10-99 nM being most potent against JAK1 (IC₅₀ = 10 nM). PF-03394197 does not inhibit a panel of 38 non-JAK kinases (IC₅₀ >1 µM). Displays activity against cytokines involved in allergy.

Formula: C₁₅H₂₃N₅O₂S
MW: 337,44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD25976611
CAS Number: 1208319-26-9
https://us.vwr.com/stibo/web/std.lang.all/23/54/23492354.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR