SB-525334, EZSolution™

Synonyms: 6-[2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl]quinoxaline6-(2-(tert-Butyl)-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl)quinoxaline

2881-5
76021-870EA 169.28 USD
76021-870
SB-525334, EZSolution™
SB-525334

A 10 mM (5 mg in 1456 µl) of the ALK5 inhibitor SB-525334 (Cat


No. 2880) in anhydrous DMSO.

Formula: C₂₁H₂₁N₅
MW: 343.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11045307
CAS Number: 356559-20-1
https://us.vwr.com/stibo/web/std.lang.all/23/52/23492352.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR