Empagliflozin ≥98% (by HPLC)

Synonyms: Jardiance(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

2877-50 2877-10
76021-864EA 461.16 USD
76021-864 76021-862
Empagliflozin ≥98% (by HPLC)
Empagliflozin

Empagliflozin is a potent and selective SGLT-2 (sodium glucose co-transporter-2) inhibitor (IC₅₀ of 3.1 nM); displays >300-fold selectivity over SGLT-1, 4, 5 and 6. Antidiabetic agent.

Formula: C₂₃H₂₇ClO₇
MW: 450.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22566222
CAS Number: 864070-44-0
https://us.vwr.com/stibo/web/std.lang.all/23/48/23492348.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Appearance Off-White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR