SBE13 hydrochloride

2847-50 2847-10
76021-782EA 145.93 USD
76021-782 76021-784
SBE13 hydrochloride
SBE13 hydrochloride

SBE13 HCl is a potent and selective PLK1 (Polo-like kinase 1) inhibitor (IC₅₀ = 200 pM)


Displays activity against PLK2 and PLK3 at higher concentrations (IC₅₀ values of 66 µM and 875 nM, respectively). Has no effect on Aurora kinase A.

Storage Temperature: Freezer
MDL Number: MFCD20036266
CAS Number: 1052532-15-6
https://us.vwr.com/stibo/web/std.lang.all/23/15/23492315.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR