TTNPB

Synonyms: (E)-4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)-1-propenyl]benzoic acid(E)-4-(2-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)prop-1-en-1-yl)benzoic acidArotinoid acid

2776-5 2776-25
76021-722EA 157.61 USD
76021-722 76021-724
TTNPB
TTNPB

TTNPB is a highly potent retinoic acid receptor (RAR) agonist (EC₅₀ = 21 nM, 4.0 nM, and 2.4 nM for RARα, RARβ, and RARγ, respectively)


Enhances reprogramming efficiency in chemically induced pluripotent stem cells (CiPSCs).

Formula: C₂₄H₂₈O₂
MW: 348.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00673917
CAS Number: 71441-28-6
https://us.vwr.com/stibo/web/std.lang.all/22/96/23492296.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR