DY131

2775-5 2775-25
76021-720EA 449.49 USD
76021-720 76021-718
DY131
DY131

DY131 is a selective agonist at estrogen-related receptors ERRβ and ERRγ


It displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM

Formula: C₁₈H₂₁N₃O₂
MW: 311.38 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00715585
CAS Number: 95167-41-2
https://us.vwr.com/stibo/web/std.lang.all/22/95/23492295.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR