ML-324

2763-5 2763-1
76021-678EA 87.57 USD
76021-678 76021-680
ML-324
ML-324

Cell-permeable


ML-324 is a JMJD2 histone demethylase inhibitor (JMJD2 IC₅₀ = 920 nM). Represses the expression of viral (HCMV and HSV-1) immediate early genes (IC₅₀ ~ 10 µM) and abrogates infection. Represses the reactivation of HSV from the latent state in sensory neurons.

Storage Temperature: Freezer
MDL Number: MFCD28053518
CAS Number: 1222800-79-5
https://us.vwr.com/stibo/web/std.lang.all/22/83/23492283.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR