OXA-01

2695-25 2695-5
76021-490EA 145.93 USD
76021-490 76021-492
OXA-01
OXA-01

OXA-01 is an ATP-competitive, selective dual inhibitor of mTORC1/mTORC2


Displays antiangiogenic and antitumor properties.

Storage Temperature: Freezer
CAS Number: 936889-68-8
https://us.vwr.com/stibo/web/std.lang.all/22/23/23492223.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR