Roflumilast

Synonyms: 3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide

2675-50 2675-10
76021-426EA 99.24 USD
76021-426 76021-428
Roflumilast
Roflumilast

Roflumilast is a selective phosphodiesterase 4 (PDE4) inhibitor (IC₅₀ = 0.8 nM)


Displays anti-inflammatory properties.

Formula: C₁₇H₁₄Cl₂F₂N₂O₃
MW: 403.21 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00938270
CAS Number: 162401-32-3
https://us.vwr.com/stibo/web/std.lang.all/22/02/23492202.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR