WHI-P154 ≥98% (by HPLC)

Synonyms: 2-Bromo-4-[(6,7-dimethoxy-4-quinazolinyl)amino]phenol2-Bromo-4-((6,7-dimethoxyquinazolin-4-yl)amino)phenol

2659-5 2659-25
76021-396EA 87.57 USD
76021-396 76021-398
WHI-P154 ≥98% (by HPLC)
WHI-P154

WHI-P154 is a potent JAK3 inhibitor (IC₅₀ of 1.8 μM). It displays no activity against JAK1 or JAK2. Also inhibits EGFR, Src, Abl, VEGFR and MAPK, and prevents Stat3, but not Stat5 phosphorylation.  Induces differentiation of neural progenitor cells


  • Appearance: White to off-white solid

Formula: C₁₆H₁₄BrN₃O₃
MW: 376.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02179366
CAS Number: 211555-04-3
https://us.vwr.com/stibo/web/std.lang.all/21/87/23492187.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR