FH535

Synonyms: 2,5-Dichloro-N-(2-methyl-4-nitrophenyl)benzenesulphonamide2,5-Dichloro-N-(2-methyl-4-nitrophenyl)benzenesulfonamide

2657-25 2657-5
76021-390EA 391 USD
76021-390 76021-388
FH535
FH535

FH535 is a potent, reversible dual inhibitor of Wnt/β-catenin and a PPARγ and PPARδ signaling


FH535 antagonizes both PPARg and PPARδ ligand-dependent activation and shows structural similarity to GW9662, a known PPARgamma antagonist. The compound is selectively toxic to some carcinomas expressing the Wnt/β-catenin pathway.

Formula: C₁₃H₁₀Cl₂N₂O₄S
MW: 361,2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01212888
CAS Number: 108409-83-2
https://us.vwr.com/stibo/web/std.lang.all/21/85/23492185.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR