MGCD0103 ≥98% (by HPLC)

Synonyms: MocetinostatMG0103N-(2-Aminophenyl)-4-[[[4-(3-pyridinyl)-2-pyrimidinyl]amino]methyl]benzamideN-(2-Aminophenyl)-4-(((4-(pyridin-3-yl)pyrimidin-2-yl)amino)methyl)benzamide

2633-10 2633-50
76021-302EA 99.24 USD
76021-302 76021-304
MGCD0103 ≥98% (by HPLC)
MGCD0103

MGCD0103 (Mocetinostat) is a class I selective HDAC inhibitor that inhibits HDAC1, HDAC2 and HDAC3 with IC₅₀ values of 0.15, 0.29 and 1.66 µM, respectively. Displays anticancer activities.


  • Appearance: White to off-white solid

Formula: C₂₃H₂₀N₆O
MW: 396.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10565970
CAS Number: 726169-73-9
https://us.vwr.com/stibo/web/std.lang.all/21/61/23492161.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR