BMS-911543 ≥98% (by HPLC)

Synonyms: BMS911543N,N-dicyclopropyl-4-((1,5-dimethyl-1H-pyrazol-3-yl)amino)-6-ethyl-1-methyl-1,6-dihydroimidazo[4,5-d]pyrrolo[2,3-b]pyridine-7-carboxamide

2630-1 2630-5
76021-290EA 99.24 USD
76021-290 76021-292
BMS-911543 ≥98% (by HPLC)
BMS-911543

BMS-911543 is a potent and selective JAK2 inhib itor. It displays potent anti-proliferative and pharmacodynamic effects in cell lines dependent upon JAK2 signaling, and has little activity in cell types dependent upon other pathways, such as JAK1 and JAK3. In addition, it displays anti-proliferative responses in colony growth assays using primary progenitor cells isolated from patients with JAK2(V617F)-positive myeloproliferative neoplasms (MPNs).


  • Appearance: Crystalline solid

Formula: C₂₃H₂₈N₈O
MW: 432.52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22200575
CAS Number: 1271022-90-2
https://us.vwr.com/stibo/web/std.lang.all/21/56/23492156.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR