STA-21

Supplier: BioVision

2626-1000 AB145648-250UG
76021-274EA 337.06 USD
76021-274 76021-276
STA-21
STA-21

STA-21 is a natural antibiotic that acts as a selective STAT3 inhibitor


It inhibits STAT3 by preventing its dimerization and DNA binding when used at concentrations of 20-30 μM. It blocks the growth and survival of cancer cells that express constitutively active STAT3 (e.g., DU145 cells, IC₅₀ = 12.2 μM). It reciprocally regulates Th17 and Treg cells, and inhibits osteoclast formation.

Storage Temperature: Freezer
CAS Number: 28882-53-3
https://us.vwr.com/stibo/web/std.lang.all/21/51/23492151.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR