Would you like to log in again?
Xestospongin C, Biovision Inc
Supplier: BioVision
2621-50 2621-10
Xestospongin C is a marine natural product isolated from Pacific basin sponges; it antagonizes the calcium-releasing action of inositol-1,4,5-trisphosphate (IP3) at the receptor level. Xestospongin C blocks the increase in intracellular calcium in vascular smooth muscle cells completely at a concentration of 25 µM. Xe C also inhibits the sarcoplasmic reticulum Ca²⁺- ATPase pump at concentrations of 700 nM.
ORDER
Expand All / Collapse All
Size Storage Temperature Package Supplier No. VWR Catalog Number Unit Price Quantity
10 µg -20 ̊C Plastic vials 2621-10 76021-254 Each (10µG) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
50 µg -20 ̊C Plastic vials 2621-50 76021-256 Each (50µG) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
CAS No. 88903-69-9
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty