UNC-669
2620-25
2620-5
76021-250EA
99.24
USD
76021-250
76021-252
UNC-669
UNC-669
UNC-669 is a small molecule nicotinamide derivative that acts asan antagonist of the MBT(Malignant Brain Tumor) domain, a family of methyl-lysine binding reader proteins
UNC669 displays selectivity for L3MBTL3 (IC₅₀ = 3.1 µM) and L3MBTL1(IC₅₀ = 4.2 µM) over L3MBTL4 (IC₅₀ = >10 µM).
Storage Temperature:
Freezer |
CAS Number:
314241-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...