Pargyline hydrochloride

Synonyms: N-Methyl-N-propargylbenzylamine Hydrochloride

2618-1000 2618-500
76021-242EA 80.52 USD
76021-242 76021-244
Pargyline hydrochloride
Pargyline hydrochloride

A selective and irreversible monoamine oxidase –B (MAO-B) inhibitor


Pargyline is also a mechanism-based inhibitor of LSD1 that effectively inhibits histone demethylation in vivo. 

Formula: C₁₁H₁₄ClN
MW: 195.6900 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012492
CAS Number: 306-07-0
https://us.vwr.com/stibo/web/std.lang.all/21/43/23492143.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR