GV-58 ≥97% (by NMR)

Warning

Synonyms: (2R)-2-[[6-[[(5-Methyl-2-thienyl)methyl]amino]-9-propyl-9H-purin-2-yl]amino]-1-butanol

2616-1 2616-5
76021-236EA 437.81 USD
76021-236 76021-234
GV-58 ≥97% (by NMR)
GV-58

GV-58 is a Roscovitine (Cat. No. 1950) derivative that acts as a selective N- and P/Q-type Ca²⁺-channel agonist. These Ca2²⁺-channels regulate transmitter release in synapses. Potential lead compound for a variety of disorders that result in neuromuscular weakness.

Formula: C₁₈H₂₆N₆OS
MW: 374.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28385865
CAS Number: 1402821-41-3
https://us.vwr.com/stibo/web/std.lang.all/21/41/23492141.jpg

Order Now

Specification Test Results

Purity 97% (NMR)
Appearance White to Off-White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR