CAL-101
Synonyms:
(S)-2-(1-((9H-Purin-6-yl)amino)propyl)-5-fluoro-3-phenylquinazolin-4(3H)-one
, Idelalisib
, GS-1101
CAL-101 is a cell-permeable inhibitor of the PI3K catalytic subunit p110δ (IC₅₀ = 2.5 nM) that displays 40- to 300-fold selectivity against other PI3K class I enzymes (IC₅₀s = 820, 565, and 89 nM for p110α, β, and γ, respectively)
It has also been shown to inhibit the chemotaxis of chronic lymphocytic leukemia cells and to downregulate the secretion of chemokines triggered by B-cell receptor signaling.
Formula:
C₂₂H₁₈FN₇O MW: 415,42 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD19443647 CAS Number: 870281-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...