Micafungin sodium

Supplier: BioVision

AB285405-5MG AB285405-25MG
76021-132EA 160.63 USD
76021-132 76021-134
Micafungin sodium
Micafungin sodium

Micafungin is an antifungal drug that belongs to the antifungal class of compounds known as echinocandins and exerts its effect by inhibiting the synthesis of 1,3-beta-D-glucan, an integral component of the fungal cell wall.

Storage Temperature: Freezer
MDL Number: MFCD08067752
CAS Number: 208538-73-2
https://us.vwr.com/stibo/web/std.lang.all/21/09/23492109.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR