CHR2797 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: alpha-[[(2R)-2-[(1S)-1-Hydroxy-2-(hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]amino]benzeneacetic acid cyclopentyl esterTosedostatCyclopentyl (S)-2-((R)-2-((S)-1-hydroxy-2-(hydroxyamino)-2-oxoethyl)-4-methylpentanamido)-2-phenylacetate

AB273387-25MG AB273387-5MG
76021-086EA 367.72 USD
76021-086 76021-088
CHR2797 ≥98% (by HPLC)
CHR2797

CHR2797 is a novel metalloenzyme inhibitor. It inhibits proliferation, and induces growth arrest and apoptosis in myeloma cells, including cells resistant to conventional chemotherapeutics.


  • Formulation: Crystalline solid

Formula: C₂₁H₃₀N₂O₆
MW: 406.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD13185162
CAS Number: 238750-77-1
https://us.vwr.com/stibo/web/std.lang.all/22/38/23492238.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR