Alisertib

Synonyms: 4-((9-Chloro-7-(2-fluoro-6-methoxyphenyl)-5H-benzo[c]pyrimido[4,5-e]azepin-2-yl)amino)-2-methoxybenzoic acidMLN8237

2718-5 2718-25
76021-080EA 758.88 USD
76021-080 76021-078
Alisertib
Alisertib

MLN8237 is a potent and selective inhibitor of Aurora kinase A (IC₅₀ =1.2 nM)


It displays >200-fold higher selectivity for Aurora kinase A over Aurora kinase B. Displays antineoplastic activity.

Formula: C₂₇H₂₀ClFN₄O₄
MW: 518,92 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16621243
CAS Number: 1028486-01-2
https://us.vwr.com/stibo/web/std.lang.all/22/36/23492236.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR