CID-5951923 ≥98% (by HPLC)

Synonyms: 2-[Methyl(tetrahydro-1,1-dioxido-3-thienyl)amino]-2-oxoethyl (E)-3-(3-nitrophenyl)prop-2-enoate

2690-5 2690-25
76021-070EA 110.91 USD
76021-070 76021-072
CID-5951923 ≥98% (by HPLC)
CID-5951923

CID-5951923 is an Inhibitor of Krüppel-like factor 5 (KLF5) transcription factor (IC₅₀ = 2.3 μM). Significantly reduces endogenous KLF5 protein levels and decreases viability of colon cancer cells, without affecting the nontransformed intestinal epithelial cells IEC-6.


  • Formulation: Off-white solid

Formula: C₁₆H₁₈N₂O₇S
MW: 382.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04796932
CAS Number: 749872-43-3
https://us.vwr.com/stibo/web/std.lang.all/22/18/23492218.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR