GSK'872
Synonyms:
N-(6-(Isopropylsulfonyl)quinolin-4-yl)benzo[d]thiazol-5-amine
, N-(6-(Isopropylsulphonyl)quinolin-4-yl)benzo[d]thiazol-5-amine
2673-25
2673-5
76021-052EA
229.14
USD
76021-052
76021-054
GSK'872
GSK'872
GSK’872 is a potent, selective, and reversible RIPK3 (receptor-interacting protein kinase-3) inhibitor.
Formula:
C₁₉H₁₇N₃O₂S₂ MW: 383,49 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD30481302 CAS Number: 1346546-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...