CID-2011756 ≥98% (by HPLC)

Synonyms: 5-(3-Chlorophenyl)-N-[4-(4-morpholinylmethyl)phenyl]-2-furancarboxamide5-(3-Chlorophenyl)-N-(4-(morpholinomethyl)phenyl)furan-2-carboxamide5-(3-Chloro-phenyl)-furan-2-carboxylic acid (4-morpholin-4-ylmethyl-phenyl)-amide

2607-25 2607-5
76021-038EA 110.91 USD
76021-038 76021-040
CID-2011756 ≥98% (by HPLC)
CID-2011756

Cell-permeable. CID-2011756 is an ATP-competitive protein kinase D (PKD) inhibitor with IC₅₀ values of 0.6, 0.7 and 3.2 μM for PKD2, PKD3 and PKD1 respectively. Inhibits phorbol ester-induced endogenous PKD1 activation in LNCaP prostate cancer cells.


  • Formulation: Off-white solid

Formula: C₂₂H₂₁ClN₂O₃
MW: 396.87 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04094273
CAS Number: 638156-11-3
https://us.vwr.com/stibo/web/std.lang.all/21/30/23492130.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR