AdipoRon ≥98% (by NMR)

Synonyms: 2-(4-Benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamideSC-3966582-(4-Benzoylphenoxy)-N-[1-(phenylmethyl)-4-piperidinyl]acetamide

2565-25 2565-5
76021-008EA 145.89 USD
76021-008 76021-010
AdipoRon ≥98% (by NMR)
AdipoRon

A potent and selective adiponectin receptor agonist (Kd = 1.8 and 3.1 μM for AdipoR1 and AdipoR2, respectively). Displays similar effects to adiponectin in muscle and liver such as activation of PPARα and AMPK signaling pathways. It also reverses insulin resistance and glucose intolerance in mice on a high-fat diet. It ameliorates diabetes in a genetically obese mouse model and prolongs the shortened lifespan of db/db mice on a high-fat diet.

Formula: C₂₇H₂₈N₂O₃
MW: 428.52 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08271060
CAS Number: 924416-43-3
https://us.vwr.com/stibo/web/std.lang.all/20/89/23492089.jpg

Order Now

Specification Test Results

Appearance White Solid
Purity ≥98% (NMR)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR