A-803467 ≥98% (by TLC)

Warning

Synonyms: 5-(4-Chlorophenyl)-N-(3,5-dimethoxyphenyl)furan-2-carboxamide

2559-5 2559-25
76020-986EA 344.42 USD
76020-986 76020-984
A-803467 ≥98% (by TLC)
A-803467

A-803467 is a potent, selective inhibitor of the TTX-resistant voltage-gated sodium channel Na(v1.8). Na(v1.8) is preferentially expressed in sensory neurons, and inhibition attenuates both inflammatory and neuropathic pain in a variety of animal models. IC50's for rat and human Na(v1.8) are 140 nM and 8.0 nM respectively.

Formula: C₁₉H₁₆ClNO₄
MW: 357.79 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10574689
CAS Number: 944261-79-4
https://us.vwr.com/stibo/web/std.lang.all/20/83/23492083.jpg

Order Now

Specification Test Results

Purity ≥98% (TLC)
Appearance Off-White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR