Evacetrapib ≥95% (by HPLC)

Synonyms: 4-trans-(((S)-5-((3,5-Bis(trifluoromethyl)benzyl)(2-methyl-2H-tetrazol-5-yl)amino)-7,9-dimethyl-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)methyl)cyclohexanecarboxylic acidtrans-4-[[(5S)-5-[[[3,5-bis(trifluoromethyl)phenyl]methyl](2-methyl-2H-tetrazol-5-yl)amino]-2,3,4,5-tetrahydro-7,9-dimethyl-1H-1-benzazepin-1-yl]methyl]- cyclohexanecarboxylic acidLY2484595

2558-25 2558-5
76020-980EA 215.99 USD
76020-980 76020-982
Evacetrapib ≥95% (by HPLC)
Evacetrapib

Evacetrapib is a potent, and selective CETP (cholesteryl ester transfer protein) inhibitor. It inhibits human recombinant CETP protein (IC₅₀ = 5.5 nM ) and CETP activity in human plasma (IC₅₀ = 36 nM) in vitro. Unlike Torcetrapib (Cat. No. 2420 & 2533), Evacetrapib does not induce aldosterone or cortisol biosynthesis.


  • Formulation: Crystalline solid

Formula: C₃₁H₃₆F₆N₆O₂
MW: 638.65 g/mol
Storage Temperature: Freezer
MDL Number: MFCD23105886
CAS Number: 1186486-62-3
https://us.vwr.com/stibo/web/std.lang.all/20/82/23492082.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR