Would you like to log in again?
Moclobemide, Biovision Inc
Supplier: BioVision
9561-10
Moclobemide is a reversible, slow-binding monoamine oxidase A (MOA-A) inhibitor (IC₅₀ = 500 µM). Displays antidepressant, anxiolytic, neuroprotective and anti-ischemic effects in vivo. Blood-brain barrier permeable.
ORDER
Expand All / Collapse All
Size Storage Temperature Package Supplier No. VWR Catalog Number Unit Price Quantity
10 mg -20°C Plastic vials 9561-10 75837-318 Each (10mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
CAS No. 71320-77-9
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty