Would you like to log in again?
Elvitegravir, Biovision Inc
Supplier: BioVision
9465-25
Elvitegravir is a novel HIV-1 integrase inhibitor that blocks strand transfer by the viral enzyme. It displays an IC₅₀ of 7.2 nM in the strand transfer assay, and shows an EC₅₀ of 0.9 nM in an acute HIV-1 infection assay.
ORDER
Expand All / Collapse All
Size Storage Temperature Package Supplier No. VWR Catalog Number Unit Price Quantity
25 mg -20°C Plastic vials 9465-25 75836-926 Each (25mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
CAS No. 697761-98-1
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty