Heat Treating Furnace, Model C133/PM3T, Cress
FC133/PM3E
80701-406EA 8880.52 USD
80701-406
Heat Treating Furnace, Model C133/PM3T, Cress
Model C133/PM3T is a heat treating furnace used for treating metals with a maximum temperature of up to 2250°F.
  • Features a Watlow PM3T single set point with timer
The chamber is constructed of firebrick of the best quality. It measures 2.5” and it is backed with 1.5” of 1800°F block insulation. The furnace shell is 18 GA. sheet metal. The door opens up away from the user. There is a rope seal on the door that provides a better seal against the furnace. The door mechanism uses two heavy duty springs that keep the door safely closed or open.
Ordering information: This unit is available in 240V, 208V. 480V is an additional charge. Optional PM4 controller with double display is available; it has four files that can hold up to 10 steps each file. Contact your VWR representative for more information.