Fluid Thioglycollate Medium with Beef Extract, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M380-500G M380-500G
95021-746EA 213.29 USD
CA95021-746 95021-746
Fluid Thioglycollate Medium with Beef Extract, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of anaerobic, microaerophilic, and aerobic microorganisms and for sterility testing.
  • Microbiological culture media
  • 500 g poly bottles