Fluid Selenite Cystine Broth, Twin Pack, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M1533I-500G M1533I-500G
95021-250EA 228.73 USD
95021-250 CA95021-250
Fluid Selenite Cystine Broth, Twin Pack, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended as an enrichment medium for the isolation of Salmonellae from feces, urine, or other pathological materials.
  • Microbiological culture media
  • 500 g poly bottles