Letheen Agar, Modified, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M946-500G M946-500G
95022-978EA 231.34 USD
CA95022-978 95022-978
Letheen Agar, Modified, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For screening cosmetic products for microbial contamination.
  • Microbiological culture media
  • 500 g poly bottles