Letheen Agar with Triton X-100, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1642-500G M1642-500G
95022-870EA 377.01 USD
CA95022-870 95022-870
Letheen Agar with Triton X-100, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For screening cosmetic products for microbial contamination.
  • Microbiological culture media
  • 500 g poly bottles