Letheen Broth with Triton X-100, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1459-500G M1459-500G
95021-162EA 289.37 USD
95021-162 CA95021-162
Letheen Broth with Triton X-100, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For screening cosmetic products for microbial contamination.
  • Microbiological culture media
  • 500 g poly bottles