Lactose Peptone Broth Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1389-500G M1389-500G
95021-072EA 227.42 USD
95021-072 CA95021-072
Lactose Peptone Broth Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of coliform organisms in water.
  • Microbiological culture media
  • 500 g poly bottles